Product References
Wip1 regulates the immunomodulatory effects of murine mesenchymal stem cells in type 1 diabetes mellitus via targeting IFN-α/BST2.
Cell death discovery
Zhou N,Liu W,Zhang W,Liu Y,Li X,Wang Y,Zheng R,Zhang Y
Published figure using CD4 monoclonal antibody (Product # MCD0404) in Flow Cytometry
Fri Oct 29 00:00:00 EDT 2021
Histone deacetylase 1 and 2 are essential for normal T-cell development and genomic stability in mice.
Blood
Dovey OM,Foster CT,Conte N,Edwards SA,Edwards JM,Singh R,Vassiliou G,Bradley A,Cowley SM
Published figure using CD4 monoclonal antibody (Product # MCD0404) in Flow Cytometry
Thu Feb 21 00:00:00 EST 2013
CD4+ Foxp3+ regulatory T cells mediate Toxoplasma gondii-induced T-cell suppression through an IL-2-related mechanism but independently of IL-10.
European journal of immunology
Tenorio EP,Fernández J,Castellanos C,Olguín JE,Saavedra R
MCD0404 was used in Flow cytometry/Cell sorting to study the role of Treg cells in Toxoplasma gondii induced suppression.
Thu Dec 01 00:00:00 EST 2011
The ΔfbpA attenuated candidate vaccine from Mycobacterium tuberculosis, H37Rv primes for a stronger T-bet dependent Th1 immunity in mice.
Tuberculosis (Edinburgh, Scotland)
Roche CM,Smith A,Lindsey DR,Meher A,Schluns K,Arora A,Armitige LY,Jagannath C
Published figure using CD4 monoclonal antibody (Product # MCD0404) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2011
Th17 cells are involved in the local control of tumor progression in primary intraocular lymphoma.
PloS one
Galand C,Donnou S,Crozet L,Brunet S,Touitou V,Ouakrim H,Fridman WH,Sautès-Fridman C,Fisson S
MCD0404 was used in flow cytometry to examine Th17 cells in the microenvironment of primary intraocular B-cell lymphoma
Fri Feb 03 00:00:00 EST 2012
Dysregulated macrophage-inflammatory protein-2 expression drives illness in bacterial superinfection of influenza.
Journal of immunology (Baltimore, Md. : 1950)
Zavitz CC,Bauer CM,Gaschler GJ,Fraser KM,Strieter RM,Hogaboam CM,Stampfli MR
MCD0404 was used in flow cytometry to develop and characterize a novel model of heterologous pulmonary infection using Bordetella parapertussis and influenza virus
Mon Feb 15 00:00:00 EST 2010
Early infection termination affects number of CD8+ memory T cells and protective capacities in listeria monocytogenes-infected mice upon rechallenge.
Journal of immunology (Baltimore, Md. : 1950)
Tseng KE,Chung CY,H'ng WS,Wang SL
MCD0404 was used in flow cytometry to assess the effects of early termination of infection on primary T cell expansion, subsequent memory cell development, and protective immunity
Wed Apr 15 00:00:00 EDT 2009
Accumulation of effector CD4 T cells during type 2 immune responses is negatively regulated by Stat6.
Journal of immunology (Baltimore, Md. : 1950)
King SB,Knorn AM,Ohnmacht C,Voehringer D
MCD0404 was used in flow cytometry to investigate the effect of Stat6 in T cell expansion, survival, and recruitment to the lung
Tue Jan 15 00:00:00 EST 2008
Dectin-1 is required for beta-glucan recognition and control of fungal infection.
Nature immunology
Taylor PR,Tsoni SV,Willment JA,Dennehy KM,Rosas M,Findon H,Haynes K,Steele C,Botto M,Gordon S,Brown GD
MCD0404 was used in flow cytometry to report that dectin-1 deficiency renders mice susceptible to infection with Candida albicans
Mon Jan 01 00:00:00 EST 2007
Complement C5a anaphylatoxin is an innate determinant of dendritic cell-induced Th1 immunity to Mycobacterium bovis BCG infection in mice.
Journal of leukocyte biology
Moulton RA,Mashruwala MA,Smith AK,Lindsey DR,Wetsel RA,Haviland DL,Hunter RL,Jagannath C
Published figure using CD4 monoclonal antibody (Product # MCD0404) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2007
Characterization of early gamma interferon (IFN-gamma) expression during murine listeriosis: identification of NK1.1+ CD11c+ cells as the primary IFN-gamma-expressing cells.
Infection and immunity
Chang SR,Wang KJ,Lu YF,Yang LJ,Chen WJ,Lin YH,Chang HH,Wang SL
MCD0404 was used in flow cytometry to identify the cells that are producing gamma interferon during infection with Listeria monocytogenes
Thu Mar 01 00:00:00 EST 2007
Renal ischemia-reperfusion injury and adenosine 2A receptor-mediated tissue protection: the role of CD4+ T cells and IFN-gamma.
Journal of immunology (Baltimore, Md. : 1950)
Day YJ,Huang L,Ye H,Li L,Linden J,Okusa MD
MCD0404 was used in flow cytometry to characterize the role of CD4+ T cells and IFN-gamma and adenosine 2A receptor-mediated tissue protection and renal ischemia-reperfusion injury
Wed Mar 01 00:00:00 EST 2006
Gene therapy of experimental autoimmune thyroiditis by in vivo administration of plasmid DNA coding for Fas ligand.
Journal of immunology (Baltimore, Md. : 1950)
Batteux F,Tourneur L,Trebeden H,Charreire J,Chiocchia G
MCD0404 was used in flow cytometry to study FasL expression on thyrocytes.
Fri Jan 01 00:00:00 EST 1999