Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
Assay-dependent | - |
Neutralization (Neu) |
0.1 µg/mL | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
24822 |
Immunogen |
Recombinant human MCP-1/CCL2 expressed in E. Coli. |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
0.5 mg/mL |
Purification |
Protein A |
Storage buffer |
PBS |
Contains |
no preservative |
Storage conditions |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
RRID |
AB_1957862 |
Neutralization: Human MCP1 is tested for its ability to neutralize the chemoattractant activity of rhMCP1 for human monocytes.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-C motif chemokine 2; chemokine (C-C motif) ligand 2; H-MCP-1; HC11; MCAF; MCP-1; MGC9434; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; Monocyte chemotactic and activating factor; Monocyte chemotactic protein 1; Monocyte secretory protein JE; RP23-350G1.3; small inducible cytokine A2 (monocyte chemotactic protein 1, homologous to mouse Sig-je); small inducible cytokine subfamily A (Cys-Cys), member 2; Small-inducible cytokine A2
Gene Aliases: CCL2; GDCF-2; HC11; HSMCR30; MCAF; MCP-1; MCP1; SCYA2; SMC-CF
UniProt ID: (Human) P13500
Entrez Gene ID: (Human) 6347
Molecular Function:
cytokine
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support