Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SDSNCCWDVS STTSSEGTVS SNIRALDLAG DPTSHKTLQK CPSTEPEEKG NIVEEQPQKN TTEKLGVSAP TIPVRRRLAW DTENTSEDVQ KQPGEKEEED DNEEEGDRKT GKQAFMGEQE KLDVHEKSKA DKMKEGSDSS VSSEKGGRLP TPKLRELGGI QRTHHDLTTP AVGGAVLVSP SKMKPPALEQ RKRMTSQDCL ETSKNDFTKK ESRAVSLLTS PAAGIKTVDP LPLREDSEDN IHKFAEATLP VSKIPKYPTN PPGQLPSPPH VPSYWHPSRR IQGSLRDPEF QHNVGKARMN NLQLPQHEAF NDEDEDRLSE ISARSAASSL RAFQTLARAK KRKENFWGKT
This gene encodes a nuclear protein similar to the mouse double minute 1 protein. The mouse gene is located in double minute (DM) chromatin particles, is amplified in the mouse transformed 3T3 cell line, and the encoded protein is able to bind to p53. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: double minute 1; Mdm1 nuclear protein homolog; Mdm4 transformed 3T3 cell double minute 1 protein; Mdm4, transformed 3T3 cell double minute 1, p53 binding protein; Mouse double minute 1; nuclear protein double minute 1; Nuclear protein MDM1
Gene Aliases: Arrd2; Mdm-1; MDM1
UniProt ID: (Human) Q8TC05, (Mouse) Q9D067
Entrez Gene ID: (Human) 56890, (Mouse) 17245
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support