Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:2,500 | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:200 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Host/Isotype |
Mouse / IgG2a |
Class |
Monoclonal |
Type |
Antibody |
Clone |
6E6E2 |
Immunogen |
ME2 Fusion Protein Ag20741 |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
500 µg/mL |
Purification |
Protein A |
Storage buffer |
PBS with 50% glycerol |
Contains |
0.02% sodium azide |
Storage conditions |
-20°C |
Aliquoting is unnecessary for -20°C storage.
ME2 encodes a mitochondrial NAD-dependent malic enzyme, a homotetrameric protein, that catalyzes the oxidative decarboxylation of malate to pyruvate. It had previously been weakly linked to a syndrome known as Friedreich ataxia that has since been shown to be the result of mutation in a completely different gene. Certain single-nucleotide polymorphism haplotypes of this gene have been shown to increase the risk for idiopathic generalized epilepsy. Alternatively spliced transcript variants encoding different isoforms found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating); Malic enzyme 2; malic enzyme 2, NAD(+)-dependent, mitochondrial; NAD-dependent malic enzyme, mitochondrial; NAD-ME; pyruvic-malic carboxylase
Gene Aliases: AW120568; D030040L20Rik; ME2; NAD-ME; ODS1
UniProt ID: (Human) P23368, (Mouse) Q99KE1
Entrez Gene ID: (Human) 4200, (Mouse) 107029, (Rat) 307270
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support