Bethyl Laboratories
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse |
Host/Isotype |
Rabbit / IgG |
Class |
Recombinant Monoclonal |
Type |
Antibody |
Clone |
BLR084G |
Immunogen |
between 2127 and 2177 (C-term) |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
250 µg/mL |
Purification |
purified |
Storage buffer |
BBS with 0.1% BSA |
Contains |
0.09% sodium azide |
Storage conditions |
2-8°C |
The initiation of transcription is controlled in part by a large protein assembly known as the preinitiation complex. A component of this preinitiation complex is a 1.2 MDa protein aggregate called Mediator. This Mediator component binds with a CDK8 subcomplex which contains the protein encoded by this gene, mediator complex subunit 12 (MED12), along with MED13, CDK8 kinase, and cyclin C.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator-recruited cofactor 240 kDa component; ARC240; CAG repeat protein 45; human opposite paired; Mediator complex subunit 12; Mediator of RNA polymerase II transcription subunit 12; mediator of RNA polymerase II transcription, subunit 12 homolog; OPA-containing protein; OPA-containing protein 1; putative mediator subunit 12; Thyroid hormone receptor-associated protein complex 230 kDa component; thyroid hormone receptor-associated protein, 230 kDa subunit; Trap230; trinucleotide repeat containing 11 (THR-associated protein); trinucleotide repeat containing 11 (THR-associated protein, 230 kDa subunit); Trinucleotide repeat-containing gene 11 protein
Gene Aliases: 230kDa; ARC240; CAGH45; FGS1; HOPA; KIAA0192; MED12; MED12S; Mopa; OHDOX; OKS; OPA-1; OPA1; TNRC11; TRAP230
UniProt ID: (Human) Q93074, (Mouse) A2AGH6
Entrez Gene ID: (Human) 9968, (Mouse) 59024
Molecular Function:
general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support