Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000-1:4,000 | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:20-1:200 | - |
Immunocytochemistry (ICC/IF) |
1:50-1:500 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
METTL4 Fusion Protein Ag12093 (125-472 aa encoded by BC111020) |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
0.25 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
PBS, pH 7.3, with 50% glycerol |
Contains |
0.02% sodium azide |
Storage conditions |
-20°C |
Immunogen sequence: ISISIIGKK RKRCVVFNQG ELDAMEYHTK IRELILDGSL QLIQEGLKSG FLYPLFEKQD KGSKPITLPL DACSLSELCE MAKHLPSLNE MEHQTLQLVE EDTSVTEQDL FLRVVENNSS FTKVITLMGQ KYLLPPKSSF LLSDISCMQP LLNYRKTFDV IVIDPPWQNK SVKRSNRYSY LSPLQIKQIP IPKLAAPNCL LVTWVTNRQK HLRFIKEELY PSWSVEVVAE WHWVKITNSG EFVFPLDSPH KKPYEGLILG RVQEKTALPL RNADVNVLPI PDHKLIVSVP CTLHSHKPPL AEVLKDYIKP DGEYLELFAR NLQPGWTSWG NEVLKFQHVD YFIALESGS (125-472 aa encoded by BC111020)
METTL4 gene ontology annotations related to this gene include methyltransferase activity; nucleic acid binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Methyltransferase-like protein 4; N(6)-adenine-specific DNA methyltransferase METTL4; N(6)-adenine-specific methyltransferase METTL4; snRNA (2'-O-methyladenosine-N(6)-)-methyltransferase METTL4
Gene Aliases: 2410198H06Rik; A730091E08Rik; AV296509; HsT661; METTL4; RGD1306451
UniProt ID: (Human) Q8N3J2, (Mouse) Q3U034
Entrez Gene ID: (Human) 64863, (Mouse) 76781, (Rat) 316731
Molecular Function:
RNA metabolism protein
RNA methyltransferase
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support