Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:300-5,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
KLH conjugated synthetic peptide derived from mouse MGMT |
Conjugate |
Biotin |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein A |
Storage buffer |
TBS with 1% BSA, 50% glycerol |
Contains |
0.03% Proclin 300 |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
O6-Methylguanine-DNA Methyltransferase (MGMT) is an important DNA repair protein involved in tumor cell resistance to the cytostatic activity of chemotherapeutic alkylating agents. This protein is also effective in protecting normal cells against the genotoxic and carcinogenic effects of DNA alkylation. The alkylating drug resistance is caused by MGMT's ability to remove DNA alkyl groups introduced in the O6 position of guanine. MGMT is expressed in highly variable amounts, depending upon the cell and tissue type, species, and cellular growth characteristics. In addition, MGMT activity varies among groups of tumors and within a particular type of tumor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 0-6-methylguanine-DNA methyltransferase; 6-O-methylguanine-DNA methyltransferase; Methylated-DNA--protein-cysteine methyltransferase; MGMT; O(6)-alkylguanine-DNA alkyltransferase; O-6-methylguan; O-6-methylguanine-DNA-alkyltransferase; O6-alkylguanine-DNA alkyltransferase; O6-methylguanine-DNA methyltranferase
Gene Aliases: Mgmt
UniProt ID: (Rat) P24528
Entrez Gene ID: (Rat) 25332
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support