Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 5
Description: The 28-14-8 monoclonal antibody reacts with the mouse MHC class I, H-2D^b, and cross-reacts with H-2L^d, H-2D^q and/or H-2L^q. Binding of 28-14-8 to the alpha 3 domain of H-2L^d is not dependent on beta2-microglobulin.
Applications Reported: This 28-14-8 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This 28-14-8 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 1.0 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.
Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222-49) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-57) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.
Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8.
HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: H-2 class I histocompatibility antigen, D-B alpha chain; H-2D cell surface glycoprotein; H-2D(B); H2-DC1-beta transplantation antigen protein; H2D1; H2DB; H2L; H2LD; MHC class I antigen; MHC class I H-2Dp; MHC class I H2 antigen; MHC H2-D-q alpha-chain; Q5k protein
Gene Aliases: H-2D; H2-D; H2-D1
UniProt ID: (Mouse) P01899
Entrez Gene ID: (Mouse) 14964
Molecular Function: major histocompatibility complex protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support