Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: EFRYKLLRAG GVMVMPEGAD TVSRPSPDYP MLPTIPLSAF SDPKKTKPSH GSKDANKESS KTSKPHKVTK EHRERPRKDS ESKSSSKELE REQAKSSKDT SRKLGEGRLP KEEKAPPPKA AFKEPKMALK ETKLESTSPK GGPPPPPPPP PRASSKRPAT ADSPKPSAKK QKKSSSKGSR SAPGTSPRTS SSSSFSDKKP AKDKSSTRGE KVKAESEPRE AKKALEVEES NSEDEASFKS ESAQSSPSNS SSSSDSSSDS DFEPSQNHSQ GPLRSMVEDL QSEESDEDDS SSGEEAAGKT NPGRDSRLSF SDSESDNSAD SSLPSREPPP PQKPPPPNSK VSGRRSPESC S
Chromosome band 11q23 is the site of translocations in myeloid and lymphoid acute leukemias, pediatric leukemias, and treatment-induced secondary acute myelogenous leukemia. The translocation breakpoints cluster in a restricted region of the HRX gene resulting in chimeric genes that encode an N-terminal portion of Hrx fused to various partner proteins. Myeloid/lymphoid or mixed-lineage leukemia translocated to 1 (MLLT1) is a nuclear protein with transcriptional transactivation properties that is fused to Hrx in t(11;19) leukemias. The minimal MLLT1 sequence required for transcription activation was narrowed to the C-terminal 90 amino acids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTC-503J8.6; ENL/MLL fusion; MLL/ENL fusion protein; MLLT1/MLL fusion; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocated to, 1; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 1; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 1; Protein ENL; YEATS domain-containing protein 1
Gene Aliases: ENL; LTG19; MLLT1; YEATS1
UniProt ID: (Human) Q03111
Entrez Gene ID: (Human) 4298
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support