Product References
The involvement of oxidants and NF-κB in cytokine-induced MMP-9 synthesis by bone marrow-derived osteoprogenitor cells.
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Ben-David D,Livne E,Reznick AZ
MA5-14244 was used in western blot to study the role of inflammatory conditions in the induction of MMP-9 production by bone marrow-derived osteoprogenitor cells
Sun Jul 01 00:00:00 EDT 2012
Matrix metalloproteinase-9 in a unique proteoglycan form in avian embryonic growth plate cartilage.
Archives of biochemistry and biophysics
Patchigolla RK,Knudson W,Schmid TM
MA5-14244 was used in western blot to study a novel proteoglycan form of MMP9 isolated from avian embryonic growth plate cartilage
Sun Apr 01 00:00:00 EDT 2012
Expression of matrix metalloproteinase-13 is controlled by IL-13 via PI3K/Akt3 and PKC-δ in normal human dermal fibroblasts.
The Journal of investigative dermatology
Moriya C,Jinnin M,Yamane K,Maruo K,Muchemwa FC,Igata T,Makino T,Fukushima S,Ihn H
MA5-14244 was used in western blot to study the mechanism by which IL-13 regulates MMP-13 expression in normal human dermal fibroblasts
Tue Mar 01 00:00:00 EST 2011
Effects of olive leaf extract and its main component oleuroepin on acute ultraviolet B irradiation-induced skin changes in C57BL/6J mice.
Phytotherapy research : PTR
Sumiyoshi M,Kimura Y
MA5-14244 was used in western blot to investigate the influence of an olive leaf extract and oleuropein on skin inflammation caused by acute UVB irradiation
Thu Jul 01 00:00:00 EDT 2010
mTOR regulates the invasive properties of synovial fibroblasts in rheumatoid arthritis.
Molecular medicine (Cambridge, Mass.)
Laragione T,Gulko PS
MA5-14244 was used in western blot to investigate the role of mTOR in the invasiveness of synovial fibroblasts in rheumatoid arthritis
Wed Dec 22 00:00:00 EST 2010
UV decreases the synthesis of free fatty acids and triglycerides in the epidermis of human skin in vivo, contributing to development of skin photoaging.
Journal of dermatological science
Kim EJ,Jin XJ,Kim YK,Oh IK,Kim JE,Park CH,Chung JH
MA5-14244 was used in western blot to investigate the influence of UV exposure on fatty acid production in the epidermis of human skin
Fri Jan 01 00:00:00 EST 2010
French maritime pine bark (Pinus maritima Lam.) extract (Flavangenol) prevents chronic UVB radiation-induced skin damage and carcinogenesis in melanin-possessing hairless mice.
Photochemistry and photobiology
Kimura Y,Sumiyoshi M
MA5-14244 was used in western blot to investigate the effects of flavangenol on UVB-caused skin damage and skin tumors
Thu Jan 13 00:00:00 EST 2011
Olive leaf extract and its main component oleuropein prevent chronic ultraviolet B radiation-induced skin damage and carcinogenesis in hairless mice.
The Journal of nutrition
Kimura Y,Sumiyoshi M
MA5-14244 was used in western blot to study the protective effects of olive leaf extract and oleuropein on UVB-irradiation induced skin damage
Sun Nov 01 00:00:00 EDT 2009
Differential regulation of MMP-13 expression in two models of experimentally induced periodontal disease in rats.
Archives of oral biology
de Aquino SG,Guimaraes MR,Stach-Machado DR,da Silva JA,Spolidorio LC,Rossa C
MA5-14244 was used in western blot to study MMP-13 expression in two models of experimentally induced periodontal disease in rats
Wed Jul 01 00:00:00 EDT 2009
Ablation of matrix metalloproteinase-9 increases severity of viral myocarditis in mice.
Circulation
Cheung C,Marchant D,Walker EK,Luo Z,Zhang J,Yanagawa B,Rahmani M,Cox J,Overall C,Senior RM,Luo H,McManus BM
MA5-14244 was used in western blot to study the severity of Coxsackievirus B3-induced myocarditis in MMP-8-and MMP-9-deficient mice
Tue Mar 25 00:00:00 EDT 2008
Bone marrow-derived cells express matrix metalloproteinases and contribute to regression of liver fibrosis in mice.
Hepatology (Baltimore, Md.)
Higashiyama R,Inagaki Y,Hong YY,Kushida M,Nakao S,Niioka M,Watanabe T,Okano H,Matsuzaki Y,Shiota G,Okazaki I
MA5-14244 was used in immunocytochemistry to study the role of bone marrow-derived cells in the spontaneous regression of liver fibrosis
Mon Jan 01 00:00:00 EST 2007
Low concentrations of transforming growth factor-beta-1 induce tubulogenesis in cultured mammary epithelial cells.
BMC developmental biology
Montesano R,Carrozzino F,Soulié P
MA5-14244 was used in western blot to investigate the effect of transforming growth factor-beta-1 at low concentrations on cultured mammary epithelial cells
Thu Feb 08 00:00:00 EST 2007
Inhibition of SMAD2 expression prevents murine palatal fusion.
Developmental dynamics : an official publication of the American Association of Anatomists
Shiomi N,Cui XM,Yamamoto T,Saito T,Shuler CF
MA5-14244 was used in western blot to study the role of SMAD2 in murine palatal fusion
Sat Jul 01 00:00:00 EDT 2006
Halofuginone induces matrix metalloproteinases in rat hepatic stellate cells via activation of p38 and NFkappaB.
The Journal of biological chemistry
Popov Y,Patsenker E,Bauer M,Niedobitek E,Schulze-Krebs A,Schuppan D
MA5-14244 was used in western blot to study the mechanisms underlying the beneficial effects of the semisynthetic plant alkaloid halofuginone in experimental liver fibrosis
Fri Jun 02 00:00:00 EDT 2006
Mechanisms of the inhibitory effect of epigallocatechin-3-gallate on cultured human vascular smooth muscle cell invasion.
Arteriosclerosis, thrombosis, and vascular biology
Cheng XW,Kuzuya M,Nakamura K,Liu Z,Di Q,Hasegawa J,Iwata M,Murohara T,Yokota M,Iguchi A
MA5-14244 was used in western blot to study the mechanisms underlying the effect of epigallocatechin-3-gallate on the invasiveness of cultured human vascular smooth muscle cells
Thu Sep 01 00:00:00 EDT 2005
TGF-beta1 modulates matrix metalloproteinase-13 expression in hepatic stellate cells by complex mechanisms involving p38MAPK, PI3-kinase, AKT, and p70S6k.
American journal of physiology. Gastrointestinal and liver physiology
Lechuga CG,Hernández-Nazara ZH,Domínguez Rosales JA,Morris ER,Rincón AR,Rivas-Estilla AM,Esteban-Gamboa A,Rojkind M
MA5-14244 was used in western blot to study the pathways by which TGF-beta1 regulates MMP-13 expression in hepatic stellate cells
Mon Nov 01 00:00:00 EST 2004
Collagen dissolution by keratinocytes requires cell surface plasminogen activation and matrix metalloproteinase activity.
The Journal of biological chemistry
Netzel-Arnett S,Mitola DJ,Yamada SS,Chrysovergis K,Holmbeck K,Birkedal-Hansen H,Bugge TH
MA5-14244 was used in western blot to study the role of cell surface plasminogen activation and MMP activity in collagen dissolution by keratinocytes
Fri Nov 22 00:00:00 EST 2002