Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
The protein encoded by this gene shares similarity with the yeast Mob1 protein. Yeast Mob1 binds Mps1p, a protein kinase essential for spindle pole body duplication and mitotic checkpoint regulation. This gene is located on the opposite strand as the interferon kappa precursor (IFNK) gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MOB kinase activator 3B; MOB kinase activator-like 2B; Mob1 homolog 2b; MOB1, Mps One Binder kinase activator-like 2B; monopolar spindle 1 binding, MOB1, domain containing; Mps one binder kinase activator-like 2B
Gene Aliases: C9orf35; MOB1D; MOB3B; MOBKL2B
UniProt ID: (Human) Q86TA1
Entrez Gene ID: (Human) 79817
Molecular Function: kinase activator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support