Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
0.2-2.5 µg/mL | - |
Immunohistochemistry (IHC) |
Assay-dependent | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
2-10 µg/mL | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Synthetic peptide region of human MEOX1. |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
0.5 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
PBS with 2% sucrose |
Contains |
0.09% sodium azide |
Storage conditions |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
RRID |
AB_2281918 |
Recommended positive controls: Jurkat.
Predicted reactivity: Cow: 92%; Dog: 92%; Guinea Pig: 83%; Horse: 92%; Mouse: 83%; Pig: 92%; Rabbit: 92%; Rat: 83%.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MEOX1 belongs to a family of nonclustered, diverged homeobox genes. It may play a role in regulating growth and differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GP91-2; Homeobox protein MOX-1; Mesenchyme homeobox 1; mitogenic oxidase 1; NOH1
Gene Aliases: KFS2; MEOX1; MOX1
UniProt ID: (Human) P50221
Entrez Gene ID: (Human) 4222
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support