Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: LKMPTQSEEE KLSPSPTTED PRVRSHVSRQ AMQGMPVRNT GSPKSAVKTR QAFFLHTTYF TKFARQVCKN TLRLRQAARR PFVAINYAAI RAECKMLLNS
Plays a role in maintenance of the normal epithelial architecture through the repression of SNAI1 transcription in a histone deacetylase-dependent manner, and thus the regulation of E-cadherin levels. Contributes to transcriptional repression by BCL6. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: metastasis associated 1 family, member 3; metastasis associated family, member 3; metastasis associated gene family, member 3; Metastasis-associated protein MTA3
Gene Aliases: KIAA1266; MTA3
UniProt ID: (Human) Q9BTC8
Entrez Gene ID: (Human) 57504
Molecular Function: homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support