Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MCDFTEDQT AEFKEAFQLF DRTGDGKILY SQCGDVMRAL GQNPTNAEVL KVLGNPKSDE MNVKVLDFEH FLPMLQTVAK NKDQGTYEDY VEGLRVFDKE GNGTVMGAEI RHVLVTLGEK MTEEEVEMLV AGHEDSNGCI NYEAFVRHIL SG
Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain that is expressed in smooth muscle and non-muscle tissues. Genomic sequences representing several pseudogenes have been described and two transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 17 kDa myosin light chain; LC17; MLC-3; Myosin light chain 3; Myosin light chain A3; Myosin light chain alkali 3; myosin light chain, alkali, nonmuscle; Myosin light polypeptide 6; myosin, light chain 6, alkali, smooth muscle and non-muscle; myosin, light polypeptide 6, alkali, smooth muscle and non-muscle; non-muscle myosin light chain; Smooth muscle and nonmuscle myosin light chain alkali 6
Gene Aliases: ESMLC; LC17; LC17-GI; LC17-NM; LC17A; LC17B; MLC-3; MLC1SM; MLC3NM; MLC3SM; MYL6; Myln
UniProt ID: (Human) P60660, (Pig) P60662, (Mouse) Q60605
Entrez Gene ID: (Human) 4637, (Pig) 396807, (Mouse) 17904, (Rat) 685867
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support