Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SKLTDEEAQ HVLEVVQRDF DLRRKEEERL EALKGKIKKE SSKRELLSDT AHLNETHCAR CLQPYQLLVN SKRQCLECGL FTCKSCGRVH PEEQGWICDP CHLARVVKIG SLEWYYEHVK ARFKRFGSAK VIRSLHGRLQ GGAGPELISE ERSGDSDQTD EDGEPGSEAQ AQAQPFGSKK KRLLSVHDFD FEGDSDDSTQ PQGHSLHLSS VPEARDSPQS LTDESCSEKA (8-236 aa encoded by BC001653)
Slac2-a (for synaptotagmin-like protein (Slp) homolog lacking C2 domains-a) links Rab27A on melanosomes with Myosin VA in melanocytes. Slac2-a binds Myosin VA through a C-terminal region and GTP-bound Rab27A through its synaptotagmin-like protein homology domain (SHD), which is located near the N-terminus. The transport of pigment and cytotoxic granules in melanocytes requires the stable formation of this complex, and thus mutations in the binding domains of the three protein components may cause albinism and/or severe immune disorders.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Exophilin-3; leaden; Melanophilin; MGC2771; MGC59733; OTTMUSP00000025451; SlaC2-a; Slp homolog lacking C2 domains a; Synaptotagmin-like protein 2a
Gene Aliases: MLPH; SLAC2-A; SLAC2A
UniProt ID: (Human) Q9BV36
Entrez Gene ID: (Human) 79083, (Pig) 100049680
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support