Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product is supplied at 1 mL (100 µL/mL).^M
^M
Application Notes: IHC-P - Sections were preincubated in 0.01 mol/l citrate, pH 6, 37°C for 10 min. Tissue sections were fixed in 4% formalin and subsequently embedded in paraffin. Atherosclerotic plaques were used as a positive control.
Methylglyoxal, also called pyruvaldehyde or 2-oxopropanal, is the organic compound with the formula CH3C(O)CHO. Gaseous methylglyoxal has two carbonyl groups, an aldehyde and a ketone but in the presence of water, it exists as hydrates and oligomers. Methylglyoxal is a reduced derivative of pyruvic acid.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2 oxo propanal; 2-Oxopropanal; Methylglyoxal; Methylglyoxal (MG); MG; MG-modified protein; Pyruvaldehyde
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support