Invitrogen
PA1-75286 specifically detects Methylprednisolone.
PA1-75286 has been successfully used in ELISA applications.
The PA1-75286 immunogen is Methylprednisolone-BTG.
Methylprednisolone is a synthetic corticosteroid with anti-inflammatory and immunomodulating properties. Methylprednisolone binds to and activates specific nuclear receptors, resulting in altered gene expression and inhibition of proinflammatory cytokine production. This agent also decreases the number of circulating lymphocytes, induces cell differentiation, and stimulates apoptosis in sensitive tumor cell populations. [National Center for Biotechnology Information. PubChem Compound Database; CID=6741].
Protein Aliases: C22H30O5; Medrate; Medrol; Medrone; Urbason