Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-23537 detects Muscarinic Acetylcholine Receptor M2 in human samples.
Muscarinic cholinergic receptor 2 (CHRM2) is an Acetylcholine Receptor has been suggested to function as a presynaptic autoreceptor that inhibits acetylcholine release. The M2 receptor has also been suggested to mediate smooth muscle contractility. Alzheimer's disease (AD) involves selective loss of M2 subtype receptors in cortical and hippocampal regions of the human brain. Increase in M2 receptor heterozygosity has been associated with depression in women. The M2 receptor has been reported to be expressed in various regions of the brain including the cerebellum, cerebral cortex, hippocampus, medulla, striatum, and thalamus, and in lung and prostate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 7TM receptor; acetylcholine receptor; acetylcholine receptor, muscarinic 2; cholinergic receptor; cholinergic receptor, muscarinic 2; cholinergic receptor, muscarinic 2, isoform a; FLJ43243; M2-mAChR; m2AChR; MGC120006; MGC120007; Muscarinic acetylcholine receptor M2; muscarinic M2 receptor
Gene Aliases: CHRM2
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support