Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Specificity: Specific for the P1 adhesin of M. pneumoniae. Does not cross react with: Ureaplasma urealyticum, Mycoplasma hominis or Mycoplasma genitalium.
Best ELISA pairs (capture/conjugate): Mycoplasma pneumoniae P1 Monoclonal antibody (Product # MA5-47571)/Mycoplasma pneumoniae Polyclonal antibody (Product # MA5-33285); Mycoplasma pneumoniae P1 Monoclonal antibody (Product # MA5-47571)/Mycoplasma pneumoniae P1 Monoclonal antibody (Product # MA5-47573).
Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 µg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 ml. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Although they are the simplest and smallest self replicating bacteria, Mycoplasma infections tend to be persistent, difficult to detect, diagnose, and cure. Mycoplasmas are surface parasites of human respiratory and urogenital tracts. Mycoplasma pneumoniae is often the causative agent of community-acquired pneumonia. The affinity of the organism for respiratory epithelial cells and its ability to secrete hydrogen peroxide are a potent combination, leading to oxidative tissue damage.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: M. pneumoniae
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support