Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:3,000 | View 2 publications 2 publications |
Immunocytochemistry (ICC/IF) |
5 µg/mL | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Published species |
Mouse |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Recombinant fragment corresponding to a region within amino acids 51 and 273 of Human MYH1 |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
0.18 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
PBS, pH 7, with 20% glycerol, 1% BSA |
Contains |
0.025% Proclin 300 |
Storage conditions |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
RRID |
AB_2548939 |
Recommended positive controls: RD, mouse heart.
Predicted reactivity: Mouse (98%), Rat (97%), Xenopus laevis (81%), Dog (98%), Pig (99%), Rabbit (97%), Rhesus Monkey (100%), Bovine (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Myosin is a major contractile protein which converts chemical energy into mechanical energy through the hydrolysis of ATP. Myosin is a hexameric protein composed of a pair of myosin heavy chains (MYH) and two pairs of nonidentical light chains. Myosin heavy chains are encoded by a multigene family. In mammals at least 10 different myosin heavy chain (MYH) isoforms have been described from striated, smooth, and nonmuscle cells. These isoforms show expression that is spatially and temporally regulated during development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2A myosin heavy chain; 2B myosin heavy chain; epididymis luminal protein 71; fast 2a myosin heavy chain; inclusion body myopathy 3, autosomal dominant; MGC133384; myh4; MyHC-2a; MyHC-2b; MyHC-2x; MyHC-IIa; MyHC-IIb; MyHC-IIx/d; Myosin heavy chain 1; Myosin heavy chain 2; Myosin heavy chain 2a; Myosin heavy chain 2b; Myosin heavy chain 2x; Myosin heavy chain 4; Myosin heavy chain IIa; Myosin heavy chain IIb; Myosin heavy chain IIx/d; myosin heavy chain type IIb; myosin heavy chain type IIx; Myosin heavy chain, skeletal muscle, adult 1; Myosin heavy chain, skeletal muscle, adult 2; Myosin heavy chain, skeletal muscle, fetal; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy chain 2, skeletal muscle, adult; myosin, heavy chain 4, skeletal muscle; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin, heavy polypeptide 2, skeletal muscle, adult; myosin, heavy polypeptide 4, skeletal muscle; Myosin-1; Myosin-2; Myosin-4; skeletal muscle myosin heavy chain II-a; type 2X myosin heavy chain; type IIA myosin heavy chain
Gene Aliases: A530084A17Rik; AI506973; HEL71; IBM3; IId; IId/x; MdMs; MHC-2X/D; MHC2A; MHC2B; MHC2X/D; Minimsc; Minmus; MM; MYH-2B; MYH1; MYH2; MYH2A; MYH2B; MYH4; MYHa; MYHas8; MYHC; MyHC-2A; MyHC-2B; MyHC-2x; MyHC-2X/D; MyHC-IIA; MyHC-IIb; MyHC-IId/x; MYHC-IIX; MyHC-IIx/d; Myhs-f; Myhs-f1; Myhs-f2; MYHSA1; MYHSA2; Myhsf; Myhsf1; Myhsf2; MYPOP
UniProt ID: (Human) P12882, (Human) Q9UKX2, (Human) Q9Y623, (Mouse) Q5SX40, (Mouse) Q5SX39, (Rat) Q29RW1
Entrez Gene ID: (Human) 4619, (Human) 4620, (Human) 4622, (Mouse) 17879, (Rat) 287408, (Rat) 691644, (Mouse) 17882, (Mouse) 17884, (Rat) 360543
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support