Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: VELSVGESKF FTCTAIGEPE SIDWYNPQGE KIISTQRVVV QKEGVRSRLT IYNANIEDAG IYRCQATDAK GQTQEATVVL EIYQKLTFRE VVSPQEFKQG EDAEVVCRVS SSPAPAVSWL YHNEEVTTIS DNRFAMLANN NLQILNINKS DEGIYRCEGR VEARGEIDFR DIIVIVNVPP AISMPQKSFN ATAERGEEMT FSCRASGSPE PAISWFRNGK LIEENEKYIL KGSNTELTVR NIINSDGGPY VCRATNKAGE DEKQAFLQVF VQPHIIQLKN ETTYENGQVT LVCDAEGEPI PEITWKRAVD GFTFTEGDKS LD
NCAM2 (neural cell adhesion molecule 2) is a cell adhesion molecule belonging to the immunoglobulin superfamily. It is differentially expressed in the developing and mature olfactory epithelium and is involved in the guidance of developing olfactory sensory neurons to the glomeruli of the olfactory bulb.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fasciclin II; N-CAM-2; NCAM-2; NCAM2; OCAM; NCAM2; OCAM; transmembrane protein; neural cell adhesion molecule RB-8; olfactory axon cell adhesion molecule; R4B12 antigen; unnamed protein product
Gene Aliases: Ncam-2; NCAM21; Ocam; OCAM-GPI; RNCAM
UniProt ID: (Human) O15394, (Mouse) O35136
Entrez Gene ID: (Rabbit) 100353943, (Human) 4685, (Rat) 288280, (Mouse) 17968
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support