Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1 µg/mL | - |
Immunocytochemistry (ICC/IF) |
5 µg/mL | - |
Immunoprecipitation (IP) |
5 µg/mL | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Synthetic peptide conjugated to KLH derived from within residues 150 - 250 of Human NCBP1. |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
0.2 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
PBS, pH 7.4, with 1% BSA |
Contains |
0.02% sodium azide |
Storage conditions |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
RRID |
AB_10987223 |
This antibody is predicted to react with mouse, rat, chicken, Xenopus laevis and cynomolgus monkey based on sequence homology.
May be involved in protein trafficking from the plasma membrane to the early endosome (EE) as well as in homotypic fusion of endocytic organelles. Mediates the endocytic trafficking from early endosomes to late endosomes and lysosomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 80 kDa nuclear cap-binding protein; CBP80; MGC2087; NCBP 80 kDa subunit; nuclear cap binding protein subunit 1, 80kD; nuclear cap binding protein subunit 1, 80kDa; Nuclear cap-binding protein subunit 1; RP11-546O6.1
Gene Aliases: CBP80; NCBP; NCBP1; Sto1
UniProt ID: (Human) Q09161
Entrez Gene ID: (Human) 4686
Molecular Function:
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support