Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NDST2 is a member of theN-deacetylase/N-sulfotransferase subfamily of the sulfotransferase1 proteins. The encoded enzyme has dual functions in processing glucosamine and heparin polymers, including N-deacetylation andN-sulfation. The encoded protein may be localized to the Golgi.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [Heparan sulfate]-glucosamine N-sulfotransferase; Bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 2; Glucosaminyl N-deacetylase/N-sulfotransferase 2; glucosaminyl N-deacetylase/N-sulphotransferase 2; heparan glucosaminyl N-deacetylase/N-sulfotransferase 2; heparan N-sulfotransferase 2; Heparan sulfate N-deacetylase 2; heparan sulfate N-deacetylase/N-sulfotransferase 2; Heparan sulfate N-sulfotransferase 2; mastocytoma N-deacetylase/N-sulfotransferase; Mndns; N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 2; N-deacetylase/N-sulfotransferase 2; N-heparan sulfate sulfotransferase 2; N-HSST 2; NDST-2
Gene Aliases: HSST2; NDST2
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support