Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
NECAB1 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EF hand calcium binding protein 1; EF-hand calcium binding protein 1; EF-hand calcium-binding protein 1; N-terminal EF-hand calcium-binding protein 1; neuronal Ca(2+)-binding protein 1; neuronal calcium binding protein; Neuronal calcium-binding protein 1; Stip-1; synaptotagmin interacting protein 1; Synaptotagmin-interacting protein 1; unnamed protein product
Gene Aliases: 1700003H21Rik; EFCBP1; NECAB1; Stip; STIP-1
UniProt ID: (Human) Q8N987, (Rat) Q9ESB5, (Mouse) Q8BG18
Entrez Gene ID: (Human) 64168, (Rat) 64169, (Mouse) 69352
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support