Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NEDD4 Antibodies

          Proteintech

          NEDD4 Recombinant Rabbit Monoclonal Antibody (230478D8)

          View all (45) NEDD4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NEDD4 Recombinant Rabbit Monoclonal Antibody (230478D8)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          NEDD4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NEDD4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NEDD4 Antibody (83112-4-RR) in ICC/IF

          Immunofluorescent analysis of (-20°C Ethanol) fixed U2OS cells using NEDD4 antibody (83112-4-RR, Clone: 230478D8) at dilution of 1:200 and Multi-rAb CoraLite ® Plus 488-Goat Anti-Rabbit Recombinant Secondary Antibody (H+L) (RGAR002). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NEDD4 Antibody in Immunocytochemistry (ICC/IF)
          NEDD4 Antibody in Immunocytochemistry (ICC/IF)
          NEDD4 Antibody in Immunocytochemistry (ICC/IF)
          NEDD4 Antibody in Western Blot (WB)
          NEDD4 Antibody in Western Blot (WB)
          NEDD4 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          NEDD4 Recombinant Rabbit Monoclonal Antibody (230478D8)

          Product Details

          83112-4-RR

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:50,000
          -

          Immunocytochemistry (ICC/IF)

          1:125-1:500
          -

          Flow Cytometry (Flow)

          0.25 µg/10^6 cells in a 100 µL suspension
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293F

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          230478D8

          Immunogen

          NEDD4 fusion protein Ag16291
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: TVLEDSYRRI MGVKRADFLK ARLWIEFDGE KGLDYGGVAR EWFFLISKEM FNPYYGLFEY SATDNYTLQI NPNSGLCNED HLSYFKFIGR VAGMAVYHGK LLDGFFIRPF YKMMLHKPIT LHDMESVDSE YYNSLRWILE NDPTELDLRF IIDEELFGQT HQHELKNGGS EIVVTNKNKK EYIYLVIQWR FVNRIQKQMA AFKEGFFELI PQDLIKIFDE NELELLMCGL GDVDVNDWRE HTKYKNGYSA NHQVIQWFWK AVLMMDSEKR IRLLQFVTGT SRVPMNGFAE LYGSNGPQSF TVEQWGTPEK LPRAHTCFNR LDLPPYESFE ELWDKLQMAI ENTQGFDGVD

          Target Information

          The E3 ubiquitin-protein ligase NEDD4 is a Ubl conjugation pathway protein. It contains one HECT (E6AP-type E3 ubiquitin-protein ligase) domain and four WW domains. It is involved in the pathway leading to the degradation of VEGFR-2/KDFR, independently of its ubiquitin-ligase activity. It acts as a potential proto-oncogene that negatively regulates PTEN via ubiquitination, a paradigm analogous to that of Mdm2 and p53. It is also reported to be involved in the budding of many viruses and is the regulator of epithelial Na+ channels, which lead to arterial hypertension. NEDD4 protein is localized to the sarcolemmal region of muscle fibers. Increase in NEDD4 protein expression leads to unloaded and denervated muscles as well as a decrease in Notch1 expression. Hence it acts as down regulator of Notch-1 gene expression.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: cell proliferation-inducing gene 53 protein; HECT-type E3 ubiquitin transferase NEDD4; KIAA0093; Nedd4; neural precursor cell expressed developmentally down-regulated protein 4; neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase; neural precursor cell expressed, developmentally down-regulated gene 4; neural precursor cell expressed, developmentally down-regulated gene 4A; OTTHUMP00000217840; OTTHUMP00000217841; OTTHUMP00000217842; PIG53; receptor-potentiating factor 1; ubiquitin protein ligase; unnamed protein product

          View more View less

          Gene Aliases: E430025J12Rik; EG639396; Gm7265; KIAA0093; NEDD4-1; Nedd4a; RPF1

          View more View less

          UniProt ID: (Human) P46934, (Rat) Q62940, (Mouse) P46935

          View more View less

          Entrez Gene ID: (Human) 4734, (Rat) 25489, (Mouse) 17999

          View more View less

          Function(s)
          ubiquitin-protein transferase activity protein binding channel inhibitor activity potassium channel inhibitor activity sodium channel inhibitor activity enzyme binding protein domain specific binding beta-2 adrenergic receptor binding ionotropic glutamate receptor binding ubiquitin binding ion channel binding phosphoserine binding phosphothreonine binding ubiquitin protein ligase activity RNA polymerase binding proline-rich region binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter ubiquitin-dependent protein catabolic process protein targeting to lysosome cellular response to DNA damage stimulus lysosomal transport neuromuscular junction development regulation of cell communication negative regulation of sodium ion transport regulation of macroautophagy protein ubiquitination regulation of signaling negative regulation of vascular endothelial growth factor receptor signaling pathway neuron projection development receptor internalization receptor catabolic process cellular response to UV regulation of protein catabolic process regulation of membrane potential glucocorticoid receptor signaling pathway ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway development involved in symbiotic interaction positive regulation of protein catabolic process viral budding positive regulation of nucleocytoplasmic transport regulation of dendrite morphogenesis regulation of synapse organization progesterone receptor signaling pathway response to calcium ion positive regulation of protein kinase B signaling protein K63-linked ubiquitination negative regulation of potassium ion export across plasma membrane adaptive immune response outflow tract morphogenesis endocardial cushion development protein monoubiquitination immune response response to denervation involved in regulation of muscle adaptation T cell activation blood vessel morphogenesis regulation of transport regulation of postsynaptic neurotransmitter receptor internalization regulation of protein catabolic process at postsynapse, modulating synaptic transmission regulation protein catabolic process at postsynapse transcription from RNA polymerase II promoter innate immune response
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-q5z7x:80/100.66.77.199:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline