Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VQWQAQEVVA QARLDGHRSM NPCPLYDAQT GTLFLFFIAI PGQVTEQQQL QTRANVTRLC QVTSTDHGRT WSSPRDLTDA AIGPAYREWS TFAVGPGHCL QLNDRARSLV VPAYAYRKLH PIQRPIPSAF CFLSHDHGRT WARGHFVAQD TLECQVAEVE TGEQRVVTLN ARSHLRARVQ AQSTNDGLDF QESQLVKKLV EPPPQGCQGS VISFPSPRSG PGSPAQWLLY THPTHSWQRA DLGAYLNPRP PAPEAWSEPV LLAKGSCAYS DLQSMGTGPD GSPLFGCLYE ANDYEEIVFL MFTLKQAFPA EYLPQ
This gene belongs to a family of glycohydrolytic enzymes which remove sialic acid residues from glycoproteins and glycolipids. Expression studies in COS7 cells confirmed that this gene encodes a functional sialidase. Its cytosolic localization was demonstrated by cell fractionation experiments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytosolic sialidase; N-acetyl-alpha-neuraminidase 2; neuraminidase 2 (cytosolic sialidase); sialidase 2 (cytosolic sialidase); Sialidase-2
Gene Aliases: NEU2; SIAL2
UniProt ID: (Human) Q9Y3R4
Entrez Gene ID: (Human) 4759
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support