Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MMLNSDTMEL DLPPTHSETE SGFSDCGGGA GPDGAGPGGP GGGQARGPEP GEPGRKDLQH LSREERRRRR RATAKYRTAH ATRERIRVEA FNLAFAELRK LLPTLPPDKK LSKIEILRLA ICYISYLNHV LDV
The helix-loop-helix (HLH) proteins are a family of putative transcription factors, some of which have been shown to play an important role in growth and development of a wide variety of tissues and species. Four members of this family have been clearly implicated in tumorigenesis via their involvement in chromosomal translocations in lymphoid tumors: MYC (MIM 190080), LYL1 (MIM 151440), E2A (MIM 147141), and SCL (MIM 187040).[supplied by OMIM, Nov 2002].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHa35; Class A basic helix-loop-helix protein 35; Helix-loop-helix protein 1; HEN-1; Nescient helix loop helix 1; NSCL-1
Gene Aliases: BHLHA35; HEN1; NHLH1; NSCL; NSCL1
UniProt ID: (Human) Q02575
Entrez Gene ID: (Human) 4807
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support