Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: LWVSQPPEIR TLEGSSAFLP CSFNASQGRL AIGSVTWFRD EVVPGKEVRN GTPEFRGRLA PLASSRFLHD HQAELHIRDV RGHDASIYVC RVEVLGLGVG TGNGTRLVVE KEHPQLG
The protein encoded by this gene is a natural cytotoxicity receptor (NCR) that may aid NK cells in the lysis of tumor cells. The encoded protein interacts with CD3-zeta (CD247), a T-cell receptor. A single nucleotide polymorphism in the 5' untranslated region of this gene has been associated with mild malaria suceptibility. Three transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, May 2010].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activating natural killer receptor p30; activating NK-A1 receptor; CD337; lymphocyte antigen 117; Natural cytotoxicity triggering receptor 3; Natural killer cell p30-related protein; natural killer cell receptor; NK-p30
Gene Aliases: 1C7; CD337; LY117; MALS; NCR3; NKp30
UniProt ID: (Human) O14931
Entrez Gene ID: (Human) 259197
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support