Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:200 | - |
Immunohistochemistry (Frozen) (IHC (F)) |
1:1,200 | - |
Immunocytochemistry (ICC/IF) |
Assay-dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Host/Isotype |
Guinea pig / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Peptide corresponding to amino acid residues 385-399 of rat GluN1 (Extracellular, N-terminus) |
Conjugate |
Unconjugated |
Form |
Lyophilized |
Concentration |
0.8 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
PBS, pH 7.4, with 1% BSA |
Contains |
0.05% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2857223 |
Reconstitute antibody with 50 µL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Peptide sequence is: ETEKPRGYQM STRLK(C)
Homology: Human, mouse amino acid residues identical
NMDAR1 encodes a protein that is a critical subunit of N-methyl-D-aspartate receptors, members of the glutamate receptor channel superfamily which are heteromeric protein complexes with multiple subunits arranged to form a ligand-gated ion channel. These subunits play a key role in the plasticity of synapses, which is believed to underlie memory and learning. Cell-specific factors are thought to control expression of different isoforms, possibly contributing to the functional diversity of the subunits. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GluN1; Glun1r; glutamate [NMDA] receptor subunit zeta 1; Glutamate [NMDA] receptor subunit zeta-1; glutamate receptor; Glutamate receptor ionotropic, NMDA 1; glutamate receptor, ionotropic, N-methyl D-aspartate 1; N-methyl-D-aspartate glutamate receptor; N-methyl-D-aspartate receptor channel, subunit zeta-1; N-methyl-D-aspartate receptor subunit NR1; neurotransmitter receptor; NMD-R1; NMDA R1 receptor C1 cassette; NMDA receptor 1; NMDZ1; NMZ1; NR1C1; NR1C2; NR1C2'; NR1N1; pGluN1; RP11-350O14.1
Gene Aliases: GluN1; GluRdelta1; Glurz1; GluRzeta1; GRIN1; M100174; MRD8; NMD-R1; NMDA1; Nmdar; NMDAR1; NR1; Rgsc174
UniProt ID: (Human) Q05586, (Rat) P35439, (Mouse) P35438
Entrez Gene ID: (Human) 2902, (Rat) 24408, (Mouse) 14810
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support