Search
Search
OriGene
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 11
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
NME1 was identified because of its reduced mRNA transcript levels in highly metastatic cells. Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of 'A' (encoded by this gene) and 'B' (encoded by NME2) isoforms. Mutations in the gene have been identified in aggressive neuroblastomas. Two transcript variants encoding different isoforms have been found for this gene. Co-transcription of this gene and the neighboring downstream gene (NME2) generates naturally-occurring transcripts (NME1-NME2), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory sperm binding protein; expressed in non-metastatic cells 1; expressed in non-metastatic cells 1, protein (NM23A) (nucleoside diphosphate kinase); farnesyl diphosphate kinase NME1; Granzyme A activated DNase (GAAD); granzyme A-activated DNase; GZMA activated DNase; histidine protein kinase NME1; included; metastasis inhibition factor NM23; NDK A; NDP kinase A; NDP kinase beta; NM23H1; NM23H1B; NME1; NME1-NME2 spliced read-through transcript; non-metastatic cells 1, protein (NM23A) expressed in; non-specific serine/threonine protein kinase NME1; nucleoside diphoshate kinase 1; nucloside diphosphate kinase; putative 3'-5'-DNA exonuclease NDK1; putative granzyme A-activated DNA endonuclease; tumor metastatic process-associated protein; unnamed protein product
Gene Aliases: AWD; GAAD; NB; NBS; NDK1; NDKA; NDPK A; NDPK-A; NDPKA; NM23; NM23-C1; NM23-H1; NM23A
UniProt ID: (Dog) Q50KA9, (Human) P15531, (Rat) Q05982
Entrez Gene ID: (Dog) 480558, (Human) 4830, (Rat) 191575
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support