Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:200 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Peptide (C)KLENASWIHDPLMK, corresponding to amino acid residues 3-16 of rat NMU2 Receptor |
Conjugate |
Unconjugated |
Form |
Lyophilized |
Concentration |
0.85 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
PBS, pH 7.4, with 1% BSA |
Contains |
0.05% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2736242 |
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 µL of deionized water.
This gene encodes a protein from the G-protein coupled receptor 1 family. This protein is a receptor for neuromedin U, which is a neuropeptide that is widely distributed in the gut and central nervous system. This receptor plays an important role in the regulation of food intake and body weight.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G-protein coupled receptor FM-4; G-protein coupled receptor TGR-1; G-protein-coupled receptor FM-4; growth hormone secretagogue receptor family, member 4; Neuromedin-U receptor 2; NMU-R2
Gene Aliases: FM-4; FM4; NMU-R2; NMU2R; NMUR2; TGR-1; TGR1
UniProt ID: (Human) Q9GZQ4, (Rat) Q9ESQ4, (Mouse) Q8BZ39
Entrez Gene ID: (Human) 56923, (Rat) 64042, (Mouse) 216749
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support