Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
The protein encoded by this gene belongs to the natriuretic peptide family. Natriuretic peptides are implicated in the control of extracellular fluid volume and electrolyte homeostasis. This protein is synthesized as a large precursor (containing a signal peptide), which is processed to release a peptide from the N-terminus with similarity to vasoactive peptide, cardiodilatin, and another peptide from the C-terminus with natriuretic-diuretic activity. Mutations in this gene have been associated with atrial fibrillation familial type 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-atrial natriuretic peptide; Alpha-hANP; ANF; ANF 1-33; ANF 3-33; ANF 8-33; ANP; Atrial natriuretic factor; Atrial natriuretic factor 1-33; Atrial natriuretic factor 3-33; Atrial natriuretic factor 8-33; Atrial natriuretic factor prohormone; Atrial natriuretic peptide; Atrial natriuretic peptide prohormone; Atriopeptigen; atriopeptin; Atriopeptin I; Atriopeptin II; Atriopeptin III; Atriopeptin-1; Atriopeptin-2; Atriopeptin-3; Auriculin-A; Auriculin-B; Auriculin-C; Auriculin-D; Cardiodilatin; cardiodilatin-related peptide; Cardionatrin; CDD; CDD 95-126; CDD-ANF; CDD-ANP (95-126); CDD-ANP (99-126); Kaliuretic peptide; KP; LANH; LANP; Long-acting natriuretic hormone; Long-acting natriuretic peptide; natriuretic peptide precursor A variant 1; Natriuretic peptides A; preproANP; preproCDD-ANF; prepronatriodilatin; Pro atrial natriuretic factor 1-30; Pro atrial natriuretic factor 31-67; Pro atrial natriuretic factor 79-98; Pro atrial natriuretic factor 99-126; Pro atrial natriuretic peptide 1-30; Pro atrial natriuretic peptide 31-67; Pro atrial natriuretic peptide 79-98; Pro atrial natriuretic peptide 95-126; proANF; proANF 1-30; proANF 31-67; proANF 79-98; proANF 99-126; proANP 1-30; proANP 31-67; proANP 79-98; proANP 95-126; Pronatriodilatin; URO; Urodilatin; Vessel dilator; VSDL
Gene Aliases: ANF; ANP; ATFB6; ATRST2; CDD; CDD-ANF; CDP; NPPA; PND
UniProt ID: (Human) P01160
Entrez Gene ID: (Human) 4878
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support