Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Positive Samples: A-549, Mouse lung; Cellular Location: Membrane, Single-pass type I membrane protein
This gene encodes one of three natriuretic peptide receptors. Natriutetic peptides are small peptides which regulate blood volume and pressure, pulmonary hypertension, and cardiac function as well as some metabolic and growth processes. The product of this gene encodes a natriuretic peptide receptor responsible for clearing circulating and extracellular natriuretic peptides through endocytosis of the receptor. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A NPR-C; ANP-C; ANPR-C; ANPRC; atrial natriuretic peptide C-type receptor; Atrial natriuretic peptide clearance receptor; Atrial natriuretic peptide clearence receptor 3; Atrial natriuretic peptide receptor 3; Atrial natriuretic peptide receptor type C; atrionatriuretic peptide receptor C; EF-2; guanylate cyclase C; longjohn; natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptide receptor C); Nppc receptor; NPR-C; strigosus; unnamed protein product
Gene Aliases: ANP-C; ANP-CR; ANPR-C; ANPRC; BOMOS; C5orf23; EF-2; GUCY2B; lgj; NPR-C; NPR3; NPRC; stri
UniProt ID: (Human) P17342, (Rat) P41740, (Mouse) P70180
Entrez Gene ID: (Human) 4883, (Rat) 25339, (Mouse) 18162
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support