Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: NGDNTDSQKL NKIISEIQAL QEVVARFRQL RLDATEFACL KCIVTFKAVP THSGSELRSF RNAAAIAALQ DEAQLTLNSY IHTRYPTQPC RFGK
Nuclear receptor TLX, a NR2 hepatocyte NF4-like receptor, has been shown to affect anterior brain differentiation, retinal development, and vision. It has been suggested that TLX targets genes for RAR beta2 and Pax2 during retinal development. Homozygous TLX mutant mice are viable at birth but exhibit reduced forebrain development and are more aggressive than wild-type mice. Mice with deleted TLX locus exhibit cerebral hypoplasia, blindness, and extreme aggression (fierce (frc) phenotype). TLX expression has been documented in mouse brain and eye. ESTs have been isolated from normal human brain libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HTll; Nuclear receptor subfamily 2 group E member 1; nuclear receptor subfamily 2, group E, member 1; Nuclear receptor TLX; Protein tailless homolog; tailes-related receptor; Tll
Gene Aliases: NR2E1; TLL; TLX; XTLL
UniProt ID: (Human) Q9Y466
Entrez Gene ID: (Human) 7101
Molecular Function: C4 zinc finger nuclear receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support