Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NOR-1, a NR4 nerve growth factor-like receptor, has been shown to affect neuronal development, T-cell apoptosis, and cancer development. NOR-1 binds to NBRE (NGFI-B response element) and regulates the pro-opiomelanocortin (POMC) gene. NOR-1 is induced rapidly but transiently by a variety of stimuli. Downregulation of NOR-1 and other NGFI-B family members is correlated with drug addiction-prone behavior of Lewis rats. Three human protein isoforms have been documented: NOR-1, NOR-1beta, and NOR-1 3' variant.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chondrosarcoma, extraskeletal myxoid, fused to EWS; Mitogen-induced nuclear orphan receptor; Neuron-derived orphan receptor 1; Nuclear hormone receptor NOR-1; Nuclear receptor subfamily 4 group A member 3; nuclear receptor subfamily 4, group A, member 3; TEC receptor; translocated in extraskeletal chondrosarcoma
Gene Aliases: CHN; CSMF; MINOR; NOR1; NR4A3; TEC
UniProt ID: (Human) Q92570
Entrez Gene ID: (Human) 8013
Molecular Function: C4 zinc finger nuclear receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support