Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NRG1 Isoform SMDF Antibodies

          Proteintech

          NRG1, isoform SMDF Recombinant Rabbit Monoclonal Antibody (240184G2)

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NRG1, isoform SMDF Recombinant Rabbit Monoclonal Antibody (240184G2)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          NRG1, isoform SMDF Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NRG1, isoform SMDF Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NRG1, isoform SMDF Antibody (83323-6-RR) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed MCF-7 cells using NRG1, isoform SMDF antibody (83323-6-RR, Clone: 240184G2) at dilution of 1:100 and CoraLite®488-Conjugated Goat Anti-Mouse IgG (H+L) (SA00013-1). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NRG1, isoform SMDF Antibody in Immunocytochemistry (ICC/IF)
          NRG1, isoform SMDF Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NRG1, isoform SMDF Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NRG1, isoform SMDF Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          NRG1, isoform SMDF Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          NRG1, isoform SMDF Antibody in Surface plasmon resonance (SPR)
          NRG1, isoform SMDF Recombinant Rabbit Monoclonal Antibody (240184G2)

          Product Details

          83323-6-RR

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:125-1:500
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          1:250
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -

          ELISA (ELISA)

          Assay-dependent
          -

          Surface plasmon resonance (SPR)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          CHO cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          240184G2

          Immunogen

          NRG1, isoform SMDF fusion protein Ag0803
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.3 mg/mL

          Amount

          30 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: MEIYSPDMSE VAAERSSSPS TQLSADPSLD GLPAAEDMPE PQTEDGRTPG LVGLAVPCCA CLEAERLRGC LNSEKICIVP ILACLVSLCL CIAGLKWVFV DKIFEYDSPT HLDPGGLGQD PIISLDATAA SALWVSSEAY TSPVSRAQSE SEVQVTVQGD KAVVSFEPSA APTPKNRIFA FSFLPSTAPS FPSPTRNPEV RTPKSATQPQ TTETNLQTAP KLSTS

          Target Information

          Neuregulin-1 (NRG1) is a gene encoding multiple isoforms involved in various physiological functions, particularly within the nervous system. The SMDF (sensory and motor neuron-derived factor) is one such isoform of NRG1. Located on chromosome 8p12, NRG1 comprises several exons, where alternative splicing gives rise to the diverse isoforms. The SMDF isoform of NRG1 plays a critical role in neural development and the maintenance of neuronal functions, including neurotransmission and synaptic plasticity. This isoform signals through interactions with ErbB family receptors, predominantly ErbB3 and ErbB4, facilitating cellular communication necessary for proper neural function. NRG1, including its SMDF variant, has been implicated in neuropsychiatric conditions such as schizophrenia and bipolar disorder, underscoring its significance in both development and disease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: acetylcholine receptor-inducing activity; glial growth factor 2; growth factor; heregulin; heregulin, alpha (45kD, ERBB2 p185-activator); neu differentiation factor; neuregulin 1; neuregulin 1 type III beta 3; NRG1; NRG1 class VII isoform alpha 2a; NRG1 class VII isoform alpha 2b; NRG1 class VII isoform alpha beta 2a; NRG1 class VII isoform alpha beta 3; NRG1 class VII isoform beta 2a; NRG1 class VII isoform beta 3; Pro-neuregulin-1, membrane-bound isoform; Pro-NRG1; sensory and motor neuron derived factor; type I neuregulin 1; type III neuregulin 1; unnamed protein product

          View more View less

          Gene Aliases: 6030402G23Rik; ARIA; D230005F13Rik; GGF; GGF2; GGFII; HGL; HRG; HRG1; HRGA; HRGalpha; MST131; MSTP131; NDF; NRG1; NRG1-IT2; SMDF

          View more View less

          UniProt ID: (Human) Q02297, (Mouse) Q6DR98

          View more View less

          Entrez Gene ID: (Human) 3084, (Rat) 112400, (Mouse) 211323

          View more View less

          Function(s)
          transcription cofactor activity receptor binding cytokine activity integrin binding protein binding growth factor activity zinc ion binding nickel cation binding protein tyrosine kinase activator activity transmembrane receptor protein tyrosine kinase activator activity receptor tyrosine kinase binding ErbB-3 class receptor binding chemorepellent activity receptor agonist activity ErbB-2 class receptor binding growth factor
          Process(es)
          ventricular trabecula myocardium morphogenesis cell communication transmembrane receptor protein tyrosine kinase signaling pathway nervous system development brain development peripheral nervous system development cell proliferation positive regulation of cell proliferation regulation of cell communication negative regulation of cardiac muscle cell apoptotic process neural crest cell development regulation of signaling cell differentiation positive regulation of cell growth mammary gland development positive regulation of protein complex assembly activation of protein kinase B activity intracellular signal transduction ERBB signaling pathway ERBB2 signaling pathway ERBB3 signaling pathway ERBB4 signaling pathway ERBB2-ERBB3 signaling pathway ERBB2-ERBB4 signaling pathway ERBB4-ERBB4 signaling pathway wound healing positive regulation of cell adhesion negative regulation of transcription, DNA-templated negative chemotaxis negative regulation of secretion positive regulation of developmental process positive regulation of striated muscle cell differentiation cardiac muscle cell differentiation ventricular cardiac muscle cell differentiation positive regulation of cardiac muscle cell proliferation cardiac muscle cell myoblast differentiation endocardial cell differentiation regulation of biological quality positive regulation of ERK1 and ERK2 cascade regulation of postsynaptic neurotransmitter receptor internalization positive regulation of peptidyl-tyrosine autophosphorylation negative regulation of extrinsic apoptotic signaling pathway in absence of ligand MAPK cascade cell morphogenesis cardiac conduction system development activation of transmembrane receptor protein tyrosine kinase activity synapse assembly heart development muscle organ development locomotory behavior regulation of blood pressure glial cell differentiation glucose transport cell migration glial cell fate commitment negative regulation of protein catabolic process myelination regulation of protein heterodimerization activity cellular protein complex disassembly neurotransmitter receptor metabolic process regulation of cell differentiation positive regulation of protein kinase activity positive regulation of Ras protein signal transduction animal organ development neuron fate commitment positive regulation of protein kinase B signaling positive regulation of protein tyrosine kinase activity positive regulation of calcineurin-NFAT signaling cascade positive regulation of cardiac muscle cell differentiation axon ensheathment positive regulation of myelination oligodendrocyte differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline