Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Immunogen sequence: MTQQPQDDFD RSVEDAQAWM KAVQDQLQVN DNTQGPRAAL EARLWETEKI CQLEPEGRVR VDLVLRMAEA LLACCPGDQK PGILARLKDI KAQWEETVTY MTHCHSRIEW VWLHWSEYLL ARDEFYRWFQ KMMVTLEPHI ELQLGLKEKQ WQLSHAQVLL HNVDNQAVLL DRLLEEAASL FNRIGDPSVD EDAQKRMKAE YDAVKAKAQK RVDLLEQVAR EHEEYQAGVD EFQLWLKAVV EKVNGCLGRN CKLPITQRLS TLQDIAKDFP RGEESLETLE EQSAGVIRNT SPLGAEKITG
As a component of the LINC (LInker of Nucleoskeleton and Cytoskeleton) complex involved in the connection between the nuclear lamina and the cytoskeleton. The nucleocytoplasmic interactions established by the LINC complex play an important role in the transmission of mechanical forces across the nuclear envelope and in nuclear movement and positioning. Probable anchoring protein which tethers the nucleus to the cytoskeleton by binding PLEC which can associate with the intermediate filament system. Plays a role in the regulation of aortic epithelial cell morphology, and is required for flow-induced centrosome polarization and directional migration in aortic endothelial cells. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KASH domain-containing protein 3; KASH3; Nesprin-3; Nuclear envelope spectrin repeat protein 3; spectrin repeat containing, nuclear envelope family member 3
Gene Aliases: C14orf139; C14orf49; LINC00341; Nesp3; nesprin-3; NET53; RGD1304563; SYNE3
UniProt ID: (Human) Q6ZMZ3, (Mouse) Q4FZC9
Entrez Gene ID: (Human) 161176, (Rat) 299356, (Mouse) 212073
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support