Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
Assay-dependent | View 1 publication 1 publication |
Immunohistochemistry (IHC) |
Assay-dependent | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:20-1:200 | - |
Immunohistochemistry (Frozen) (IHC (F)) |
1:20-1:200 | - |
Immunocytochemistry (ICC/IF) |
1:20-1:200 | View 1 publication 1 publication |
Product Specifications | |
---|---|
Species Reactivity |
Rat, Mouse |
Published species |
Mouse |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
2Q178 |
Immunogen |
Full length native Nestin protein purified from embryonic rat spinal cord. |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein A |
Storage buffer |
0.1M tris glycine with 0.15M NaCl |
Contains |
0.05% sodium azide |
Storage conditions |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
RRID |
AB_2151007 |
For use in ICC applications, cells can be fixed with 4% paraformaldehyde buffered with 50mM sodium borate at p.H. 9.5. for IHC-P and IHC-Fr tissue can be fixed with 4% paraformaldehyde at pH 7.4 for light microscopy.
The predicted molecular weight of MA1-91657 is ~200kDa.
Nestin is a Class VI intermediate filament abundantly produced in the developing central nervous system (CNS) and in early embryonic neuroepithelial stem cells. Nestin has been widely used as a predominant marker for stem/progenitor cells, glioma cells, and tumor endothelial cells. Also, Nestin is a superior angiogenic marker to evaluate neovascularity of endothelial cells in tumors. Highly specific antibodies to human Nestin can aid in characterizing progenitor cells differentiating into distinct lineages, in enhancing the therapeutic potential of human neural stem/progenitor cells in the treatment of CNS diseases or injury, and in identifying neuroepithelial tumor cells. Nestin expression has been observed in pancreatic hematopoietic stem cell populations, and is considered a novel angiogenesis marker of proliferating endothelial cells in colorectal cancer tissue.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ21841; intermediate filament protein; Nestin
Gene Aliases: AA166324; C78523; ESTM46; Nes
UniProt ID: (Mouse) Q6P5H2
Entrez Gene ID: (Rat) 25491, (Mouse) 18008
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support