Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Neuroligin 1 Antibodies

          Antibodies Inc

          Neuroligin-1 Monoclonal Antibody (N97A/31)

          View all (36) Neuroligin 1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Neuroligin-1 Monoclonal Antibody (N97A/31)

          • Antibody Testing Data (4)
          Neuroligin-1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Neuroligin-1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Neuroligin-1 Antibody (75-160) in ICC/IF

          Confocal microscopy analysis of NLGN1(red) and APC (magenta) co-staining in HuTu 80 cells. Image from publication CC-BY-4.0. PMID:36056393. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Neuroligin-1 Antibody in Immunocytochemistry (ICC/IF)
          Neuroligin-1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Neuroligin-1 Antibody in Western Blot (WB)
          Neuroligin-1 Antibody in Western Blot (WB)
          Neuroligin-1 Monoclonal Antibody (N97A/31)

          Product Details

          75-160

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          1:500
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Chicken, Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N97A/31

          Immunogen

          Fusion protein amino acids 718-843 (intracellular C-terminus) of rat Neuroligin-1 (accession number NP_446320) produced recombinantly in E. Coli.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4

          Contains

          0.065% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          Product Specific Information

          For maximum recovery of product, centrifuge the vial prior to removing the cap.

          Target Information

          This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: MGC45115; neuroligin 1 isoform A1A2B; Neuroligin I; Neuroligin-1; NLG 1

          View more View less

          Gene Aliases: 6330415N05Rik; BB179718; KIAA1070; mKIAA1070; neuroligin-1; NL1; Nlg1; NLGN1

          View more View less

          UniProt ID: (Human) Q8N2Q7, (Mouse) Q99K10, (Rat) Q62765

          View more View less

          Entrez Gene ID: (Human) 22871, (Chicken) 424980, (Mouse) 192167, (Rat) 116647

          View more View less

          Function(s)
          receptor activity PDZ domain binding neurexin family protein binding protein dimerization activity cell adhesion molecule binding carboxylic ester hydrolase activity scaffold protein binding protein binding cell adhesion molecule
          Process(es)
          regulation of respiratory gaseous exchange by neurological system process protein targeting heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules neuron cell-cell adhesion nervous system development synapse assembly positive regulation of circadian sleep/wake cycle, wakefulness synaptic vesicle targeting calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules neuronal signal transduction neuron projection development positive regulation of synaptic transmission, GABAergic protein localization to synapse establishment of protein localization regulation of neuron differentiation rhythmic process cytoskeletal matrix organization at active zone protein homooligomerization protein heterotetramerization positive regulation of filopodium assembly positive regulation of synapse assembly positive regulation of synaptic transmission, glutamatergic long-term synaptic potentiation positive regulation of dendritic spine development negative regulation of dendritic spine morphogenesis terminal button organization synaptic vesicle clustering postsynaptic membrane assembly presynaptic membrane assembly AMPA glutamate receptor clustering NMDA glutamate receptor clustering neurexin clustering involved in presynaptic membrane assembly postsynaptic density protein 95 clustering receptor localization to synapse positive regulation of ruffle assembly positive regulation of synaptic vesicle endocytosis positive regulation of protein localization to synapse positive regulation of intracellular signal transduction positive regulation of synaptic vesicle exocytosis regulation of N-methyl-D-aspartate selective glutamate receptor activity regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity positive regulation of excitatory postsynaptic potential positive regulation of synaptic vesicle clustering cell adhesion synaptic vesicle transport modulation of synaptic transmission synapse organization inhibitory postsynaptic potential metabolic process neuronal ion channel clustering
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0