Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Neuropilin 1 (CD304) Antibodies

          Invitrogen

          Neuropilin 1 Recombinant Rabbit Monoclonal Antibody (0A7X1)

          View all (53) Neuropilin 1 (CD304) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Neuropilin 1 Recombinant Rabbit Monoclonal Antibody (0A7X1)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (11)
          Neuropilin 1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Neuropilin 1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Neuropilin 1 Antibody (MA569506) in ICC/IF

          Immunocytochemical analysis of Neuropilin 1 in Neuro-2a cells. Samples were incubated with Neuropilin 1 recombinant monoclonal antibody (Product # MA569506) using a dilution of 1:200 (40x lens). Followed by a further incubation with Cy3 Goat Anti-Rabbit IgG (H+L) using a dilution of 1:500 (Red). The cells were counterstained with alpha-Tubulin Mouse mAb using dilution of 1:400. Followed by incubation with 4... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Neuropilin 1 Antibody in Immunocytochemistry (ICC/IF)
          Neuropilin 1 Antibody in Immunocytochemistry (ICC/IF)
          Neuropilin 1 Antibody in Immunocytochemistry (ICC/IF)
          Neuropilin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Neuropilin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Neuropilin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Neuropilin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Neuropilin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Neuropilin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Neuropilin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Neuropilin 1 Antibody in Western Blot (WB)

          Product Details

          MA569506

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:700-1:7,000
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          0A7X1

          Immunogen

          Recombinant protein (or fragment).

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.32 mg/mL

          Amount

          132 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.3., with 50% glycerol

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          Neuropilin 1 is a 923 amino acid containing membrane bound coreceptor to a tyrosine kinase receptor belonging to the neuropilin family with two CUB domains, two F5/8 type C domains and a MAM domain. Neuropilin 1 forms a heterodimer with NRP2 (Neuropilin 2) and is known to bind to plexin (PLXNB1) to form semaphorin receptors. Expressed mostly in the nervous system this receptor is implicated in a wide range of functions including in the development of the cardiovascular system, angiogenesis, repulsive axon guidance, formation of certain neuronal circuits, cell survival, control of neuronal migration and organogenesis outside the nervous system. This semaphorin receptor functions as receptors for both the semaphorins and vascular endothelial growth factor and is also a regulator of VEGF-induced angiogenesis. Neuropilin 1 may be involved in aggressive advanced prostate carcinoma and in the progression of breast cancer.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: A5 protein; alternatively spliced; lacks exon 16; CD304; Neuropilin; Neuropilin-1; Neuropilin-1 precursor (A5 protein); Neuropilin1; NRP 1; receptor for VEGF165 and semaphorins class 3; sNRP 1; sNRP1; soluble NRP 1; soluble NRP1; transmembrane receptor; unnamed protein product; Vascular endothelial cell growth factor 165 receptor; VEGF 165 receptor

          View more View less

          Gene Aliases: BDCA4; C530029I03; CD304; NP-1; NP1; NPN-1; Npn1; NRP; NRP1; VEGF165R

          View more View less

          UniProt ID: (Human) O14786, (Rat) Q9QWJ9, (Mouse) P97333

          View more View less

          Entrez Gene ID: (Human) 8829, (Rat) 246331, (Mouse) 18186

          View more View less

          Function(s)
          vascular endothelial growth factor-activated receptor activity GTPase activator activity protein binding heparin binding coreceptor activity semaphorin receptor activity growth factor binding protein kinase binding cytokine binding vascular endothelial growth factor binding metal ion binding oxidoreductase
          Process(es)
          angiogenesis patterning of blood vessels vasculogenesis neural crest cell migration neuron migration positive regulation of endothelial cell proliferation sprouting angiogenesis cell migration involved in sprouting angiogenesis outflow tract septum morphogenesis substrate-dependent cell migration, cell extension signal transduction integrin-mediated signaling pathway cell-cell signaling nervous system development axon guidance axonal fasciculation heart development motor neuron axon guidance response to wounding organ morphogenesis positive regulation of endothelial cell migration positive regulation of smooth muscle cell migration dendrite development facial nerve structural organization trigeminal nerve morphogenesis trigeminal nerve structural organization vestibulocochlear nerve structural organization branchiomotor neuron axon guidance gonadotrophin-releasing hormone neuronal migration to the hypothalamus cell differentiation negative regulation of axon extension regulation of vascular endothelial growth factor receptor signaling pathway retinal ganglion cell axon guidance regulation of Cdc42 protein signal transduction cellular response to hepatocyte growth factor stimulus endothelial cell chemotaxis cellular response to vascular endothelial growth factor stimulus ventral trunk neural crest cell migration vascular endothelial growth factor signaling pathway neuropilin signaling pathway VEGF-activated neuropilin signaling pathway positive regulation of phosphorylation negative regulation of neuron apoptotic process blood vessel endothelial cell migration endothelial cell migration positive regulation of angiogenesis viral entry into host cell platelet-derived growth factor receptor signaling pathway vascular endothelial growth factor receptor signaling pathway hepatocyte growth factor receptor signaling pathway sympathetic nervous system development neuron development axon extension regulation of axon extension involved in axon guidance positive regulation of axon extension involved in axon guidance negative regulation of axon extension involved in axon guidance artery morphogenesis axon extension involved in axon guidance positive chemotaxis positive regulation of filopodium assembly positive regulation of stress fiber assembly positive regulation of focal adhesion assembly axonogenesis involved in innervation regulation of vesicle-mediated transport dichotomous subdivision of terminal units involved in salivary gland branching angiogenesis involved in coronary vascular morphogenesis coronary artery morphogenesis retina vasculature development in camera-type eye retina vasculature morphogenesis in camera-type eye renal artery morphogenesis sympathetic ganglion development trigeminal ganglion development positive regulation of ERK1 and ERK2 cascade semaphorin-plexin signaling pathway commissural neuron axon guidance positive regulation of cell migration involved in sprouting angiogenesis endothelial tip cell fate specification sensory neuron axon guidance motor neuron migration sympathetic neuron projection extension sympathetic neuron projection guidance postsynapse organization regulation of plasma membrane bounded cell projection organization basal dendrite development basal dendrite arborization positive regulation of substrate adhesion-dependent cell spreading neural crest cell migration involved in autonomic nervous system development protein localization to early endosome facioacoustic ganglion development dorsal root ganglion morphogenesis otic placode development negative regulation of extrinsic apoptotic signaling pathway cell migration peptidyl-tyrosine phosphorylation nerve development otic placode formation positive regulation of peptidyl-tyrosine phosphorylation ganglion morphogenesis regulation of retinal ganglion cell axon guidance toxin transport semaphorin-plexin signaling pathway involved in neuron projection guidance semaphorin-plexin signaling pathway involved in axon guidance positive regulation of retinal ganglion cell axon guidance VEGF-activated neuropilin signaling pathway involved in axon guidance multicellular organism development cell migration involved in coronary vasculogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-fcf5f9d6d-txc6f:80/100.66.78.195:80.
          git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.51.0-2026.05.21-1.0