Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSILFYVIF LAYLRGIQGN NMDQRSLPED SLNSLIIKLI QADILKNKLS KQMVDVKENY QSTLPKAEAP REPERGGPAK SAFQPVIAMD TELLRQQRRY NSPRVLLSDS TPLEPPPLYL MEDYVGSPVV ANRTSRRKRY AEHKSHRGEY SVCDSESLWV TDKSSAIDIR GHQVTVLGEI KTGNSPVKQY FYETRCKEAR PVKNGCRGID DKHWNSQCKT SQTYVRALTS ENNKLVGWRW IRIDTSCVCA LSRKIGRT (1-257 aa encoded by BC069773)
The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HDNF; Nerve growth factor 2; Neurotrophic factor; Neurotrophin-3; neurotrophin-3 (HDNF/NT-3); NGF-2; NT-3
Gene Aliases: AI316846; AI835689; HDNF; NGF-2; NGF2; NT-3; NT3; Ntf-3; NTF3
UniProt ID: (Human) P20783, (Mouse) P20181, (Rat) P18280
Entrez Gene ID: (Human) 4908, (Mouse) 18205, (Rat) 81737
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support