Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:2,000 | - |
Immunocytochemistry (ICC/IF) |
1:100 | - |
Flow Cytometry (Flow) |
1:100 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
OTI1C5 |
Immunogen |
Full length human recombinant protein of human OBFC1 produced in HEK293T cell. |
Conjugate |
Unconjugated |
Form |
liquid |
Concentration |
0.35 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS with 1% BSA, 50% glycerol |
Contains |
0.02% sodium azide |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
OBFC1 is a component of the CST complex, a complex that binds to single-stranded DNA and is required to protect telomeres from DNA degradation. The CST complex binds single-stranded DNA with high affinity in a sequence-independent manner, while isolated subunits bind DNA with low affinity by themselves. In addition to telomere protection, the CST complex has probably a more general role in DNA metabolism at non-telomeric sites.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha accessory factor 44; CST complex subunit STN1; oligonucleotide/oligosaccharide-binding fold containing 1; Oligonucleotide/oligosaccharide-binding fold-containing protein 1; replication protein A 32 kDa subunit; Suppressor of cdc thirteen homolog
Gene Aliases: AAF-44; AAF44; bA541N10.2; OBFC1; RPA-32; STN1
UniProt ID: (Human) Q9H668
Entrez Gene ID: (Human) 79991
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support