Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPARALLPR RMGHRTLAST PALWASIPCP RSELRLDLVL PSGQSFRWRE QSPAHWSGVL ADQVWTLTQT EEQLHCTVYR GDKSQASRPT PDELEAVRKY FQLDVTLAQL YHHWGSVDSH FQEVAQKFQG VRLLRQDPIE CLFSFICSSN NNIARITGMV ERLCQAFGPR LIQLDDVTYH GFPSLQALAG PEVEAHLRKL GLGYRARYVS ASARAILEEQ GGLAWLQQLR ESSYEEAHKA LCILPGVGTK VADCICLMAL DKPQAVPVDV HMWHIAQRDY SWHPTTSQAK GPSPQTNKEL GNFFRSLWGP YAGWAQAVLF SADLRQCRHA QEPPAKRRKG SKGPEG
8-hydroxyguanine, a form of oxidative DNA damage induced by free radicals, causes G:C to T:A transversion. In E. Coli, three DNA repair enzymes exist to prevent the mutagenic effects of 8-hydroxyguanine. One of these enzymes, MutM, was found to have a functional yeast (yOgg1) and human (hOgg1) homologue. hOgg1 proteins efficiently released the 8-hydroxyguanine opposite the pyrimidine from DNA and cleaved the AP site in a manner similar to bacterial and yeast enzymes. Genetic backgrounds in control of the repair of damaged DNA are involved in the susceptibility to cancer development. The hOgg1 gene has been mapped to region 3p26. 2, a region showing loss of heterozygosity (LOH) in a variety of cancers. In particular, 3p25-p26 is a common LOH region in lung cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 8-hydroxyguanine DNA glycosylase; AP lyase; DNA-apurinic or apyrimidinic site lyase; N-glycosylase/DNA lyase; OGG1 type 1f
Gene Aliases: HMMH; HOGG1; MMH; MUTM; OGG1; OGH1
UniProt ID: (Human) O15527
Entrez Gene ID: (Human) 4968
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support