Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MPGRWRWQRD MHPARKLLSL LFLILMGTEL TQNKRENKAE KMGGPESERK TTGEKTLNEL PLFCLEAHAG SLALPRMCSP NPNPAVGLCR PAYPQSPSPG AAQTISQSLL ERFCMASRRE VFLAPGRPGG GWWLCTVAGQ MHSFMCTHTH THAHTGEQIP AEKSQPGPD
The Olfactomedin family comprises a diverse group of secreted glycoproteins, which includes OLFM1 (Noelin-1), OLFM2 (Noelin-2), OLFM3 (Noelin-3), OLFM4 (Noelin-4), tiarin, pancortin, gliomedin and mycocilin. These proteins are implicated in the development of the nervous system. Specifically, OLFM1 and OLFM2 expression is observed in the neural plate and neural crest, as well as in the cranial ganglia in mouse at E8-10, and later in brain tissue and in the zone of polarizing activity in the limb. Overexpression of OLFM1 causes an excess of neural crest emigrations and prolonged neural crest production. OLFM2 participates in the regulation of the development of the anterior nervous system. An Arg144Gln mutation in OLFM2 has been implicated as a possible cause for open-angle glaucoma (OAG).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: neuroblastoma protein; Neuronal olfactomedin-related ER localized protein; Noelin; olfactomedin related ER localized protein; Olfactomedin-1; pancortin 1
Gene Aliases: AMY; NOE1; NOEL1; NOELIN1; OlfA; OLFM1
UniProt ID: (Human) Q99784
Entrez Gene ID: (Human) 10439
Molecular Function: structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support