Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MKEEAFLRRR FSLCPPSSTP QKVDPRKLTR NLLLSGDNEL YPLSPGKDME PNGPSLPRDE GPPTPSSATK VPPAEYRLCN GSDKECVSPT ARVTKKETLK AQKENYRQEK KRATRQLLSA LTDPSVVIMA DSLKGPKGES VGSITQPLPS SYLIFRAASE SDGRCWLDAL ELALRCSSLL RLGTCKPGRD GEPGTSPDAS PSSLCGLPAS ATVHPDQDLF PLNGSSLEND AFSDKSEREN PEESDTETQD HSRKTESGSD QSETPGAPVR RGTTYVEQVQ EELGELGEAS QVETVSEENK SLMWTLLKQL RPGMDLSRVV LPTFVLEPRS FLNKLSDYYY HADLLSRAAV EED
This gene encodes a member of the oxysterol-binding protein family, a group of intracellular lipid receptors that play a key role in the maintenance of cholesterol balance in the body. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. This gene has been shown to be imprinted, with preferential expression from the maternal allele only in placenta. Transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp586H1719; FLJ31948; FLJ42929; ORP-5; OSBP-related protein 5; oxysterol binding protein 2; oxysterol binding protein homolog 1; oxysterol binding protein-like 5; Oxysterol-binding protein homolog 1; oxysterol-binding protein homologue 1; oxysterol-binding protein-like protein 5; Oxysterol-binding protein-related protein 5
Gene Aliases: 1110006M06Rik; AI462538; KIAA1534; OBPH1; ORP5; Osbp2; OSBPL5
UniProt ID: (Human) Q9H0X9, (Mouse) Q9ER64
Entrez Gene ID: (Human) 114879, (Mouse) 79196
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support