Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
May be involved in meiosis or the maturation of germ cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hT41; Oxidative stress-induced growth inhibitor 2
Gene Aliases: C8orf1; hT41; OSGIN2
UniProt ID: (Human) Q9Y236
Entrez Gene ID: (Human) 734
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support