Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 6
This antibody is predicted to react with Monkey, Cat, Dog
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
P2Y12 belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is involved in platelets aggregation, and is a potential target for the treatment of thromboembolisms and other clotting disorders. Two transcript variants encoding the same isoform have been identified for this gene. Expression of P2Y12 has been reported in platelets, spinal cord, and many brain regions. ESTs for P2Y12 have been isolated from B-cell/lung/testis, brain, embryo, prostate, eye, kidney carcinoma, and colon carcinoma libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-glucose receptor; ADPG-R; G-protein coupled receptor SP1999; Gi-coupled ADP receptor HORK3; P2T(AC); P2Y purinoceptor 12; P2Y(AC); P2Y(ADP); P2Y(cyc); P2Y12; P2Y12 platelet ADP receptor; P2YR12; purinergic receptor P2RY12; purinergic receptor P2Y, G-protein coupled, 12; putative G-protein coupled receptor; SP1999
Gene Aliases: 2900079B22Rik; 4921504D23Rik; ADPG-R; BDPLT8; HORK3; P2RY12; P2T(AC); P2Y(12)R; P2Y(AC); P2Y(ADP); P2Y(cyc); P2Y12; SP1999
UniProt ID: (Human) Q9H244, (Mouse) Q9CPV9, (Rat) Q9EPX4
Entrez Gene ID: (Human) 64805, (Mouse) 70839, (Rat) 64803
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support