Search Thermo Fisher Scientific
Recommended positive controls: HepG2.
Predicted reactivity: Cow: 85%; Dog: 85%; Guinea Pig: 92%; Horse: 92%; Mouse: 83%; Rat: 83%; Sheep: 85%.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
D-site-binding protein (DBP) is a member of the PAR bZIP (proline and acidic amino acid-rich basic leucine zipper) transcription factor family.
Protein Aliases: Albumin D box-binding protein; Albumin D-element-binding protein; D site of albumin promoter (albumin D-box) binding protein; D site-binding protein; Tax-responsive enhancer element-binding protein 302; TaxREB302
Gene Aliases: DBP