Search Thermo Fisher Scientific
This antibody is predicted to react with bovine based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
IGF-BPs control the distribution, function and activity of IGFs in various cell tissues and body fluids. IGF-BP6, which specifically inhibits IGF-II actions, is produced by bone cells and is the major IGF-BP present in cerebrospinal fluid. IGF-BP6 has been shown to inhibit IGF-II-dependent cancers such as neuroblastoma, colon cancer and rhabdomyosarcoma.
Protein Aliases: IBP-6; IGF binding protein 6; IGF-binding protein 6; IGFBP; IGFBP-6; Insulin like growth factor binding protein; insulin-like growth factor binding protein 6; Insulin-like growth factor-binding protein 6
Gene Aliases: IBP6; IGFBP6
UniProt ID: (Human) P24592
Entrez Gene ID: (Human) 3489
Molecular Function: protease inhibitor