Search Thermo Fisher Scientific
May act as a scaffold to regulate the recruitment and assembly of spindle midzone components. Required for the localization of AURKB and PLK1 to the spindle midzone. [UniProt]
Protein Aliases: Coiled-coil domain-containing protein 69
Gene Aliases: CCDC69; D11Ertd461e